Learn More
Kinesis™ Adapters for HPLC
Convenience of quick connect and disconnect of a luer connection
£13.60 - £16.10
Product Code | Brand | Connections | For Use With (Equipment) | Material | Price | Quantity & Availability | |||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Connections | For Use With (Equipment) | Material | Price | Quantity & Availability | |||
12977719
|
Kinesis™
P-604 |
Female internal threads Luer to 1/4-28 Flat-bottom | Female Luer x Male 1/4-28 Flat Bottom | Acetal |
£13.60
Each |
Estimated Shipment: 30-12-2024
Log in to see stock. |
|||
12987739
|
Kinesis™
02014-06 |
Female Luer to Male 10-32 Adapter | Female Luer to Male 10-32 Adapter | ETFE |
£16.10
Each |
In Stock
Log in to see stock. |
|||
Description
Convenience of quick connect and disconnect of a luer connection
- Manufactured from inert polymers
Quick connect luer adapters are designed to connect any female luer to any male luer in a variety of applications. Use with male luer lock adapters for more secure connection.