Learn More
Kinesis™ Adapters for HPLC
Convenience of quick connect and disconnect of a luer connection
Brand:  Kinesis™ 02014-06
 View more versions of this product
Product Code. 12987739
  • £16.10 / Each
In Stock
to see stock.
Explore more special offers
Add to basket
Description
Description
Convenience of quick connect and disconnect of a luer connection
  • Manufactured from inert polymers
Quick connect luer adapters are designed to connect any female luer to any male luer in a variety of applications. Use with male luer lock adapters for more secure connection.
Specifications
Specifications
Adapter
Female Luer to Male 10-32 Adapter
Female Luer to Male 10-32 Adapter
0.05 in.
ETFE