Learn More
Kinesis™ Quick Connect Adapter
Convenience of quick connect and disconnect of a luer connection
Brand: Kinesis™ 02014-21
This item is not returnable.
View return policy
Description
Convenience of quick connect and disconnect of a luer connection
Quick connect threaded luer adapters are designed to connect any male luer to any female luer in a variety of applications. Male luer fittings are available with or without lock hubs. Lock hubs provide added security of holding the connection in place.Specifications
Male Luer Adapter | |
40°C | |
Red |
0.05 in. | |
ETFE | |
Male Luer Lock x Female 1/4-28 Flat Bottom |