missing translation for 'onlineSavingsMsg'
Learn More

Kinesis™ Shut-off Valves

Quickly stop a flow stream

Brand:  Kinesis™ 02014-00

 View more versions of this product

Product Code. 13090400

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »

Explore more special offers
This item is not returnable. View return policy

Description

Description

Quickly stop a flow stream

  • Withstands pressures up to 500 psi
  • Quickly stop a flow stream
  • All valves feature 1/4-28 flat-bottom ports and come with fittings.
Specifications

Specifications

Natural
500 psi (34 bar)
2.5 μL (Internal)
1.58 mm
PEEK
Shut-Off Valve
0.062 in.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.