missing translation for 'onlineSavingsMsg'
Learn More

Liebherr™ Upright Ultra-low Freezers

Liebherr™ -86°C Upright Ultra-low Freezers guarantee reliable, energy-efficient and safe storage validated using EN 60068-3. Robust, long-lasting and user-friendly with a wide temperature range from -40°C to-86°C from Liebherr, the refrigeration specialists.

Brand:  Liebherr™ SUFSG5001

13195.56 GBP valid until 2025-02-21
Use promo code "24115" to get your promotional price.


 View more versions of this product

Product Code. 17260587

  • £16790.00 / Each

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

Two models availible to suit your storage needs and lab footprint specifications

Performance:

  • Liebherr's refrigeration expertise ensures the 2-stage refrigeration system is well-balanced and features an array of pressure and temperature controls for reliable and long-term performance. Water-cooled versions available to avoid local heat emission
  • Multi-chamber door gasket reduces both energy consumption and condensation around the door

Sustainability:

  • Liebherr ULT freezers use only natural refrigerants compliant with the EU F-Gas directive and other sustainability standards.
  • The Liebherr ULT freezers have in-situ foamed polyurethane insulation enhanced with high-quality VIP panels for maximum energy-efficiency, low heat emission and excellent temperature stability

User benefits:

  • Clean and attractive white exterior design
  • 3 stainless steel shelves, height-adjustable in 24 mm increments, adapt to all storage configurations and allow optimum utilisation of chamber volume
  • User-friendly, intuitive controls including extensive alarm and security functions for added peace of mind. 3-stage alarm and monitoring system
  • Easy door-opening with heated pressure relief valve and heavy-duty all-metal lever door handle
  • Wide temperature range -40°C to -86°C for greater flexibility to cover a variety of user requirements
  • Clip-on removable unit cover gives easy access to the washable condenser filter for simple maintenance
  • Stainless steel drip tray incorporated in the unit cover catches water from the chamber making this part of defrosting and cleaning much easier.
  • Quiet operation and low heat-emission for greatest siting flexibility. 47 dB(A) sound power level.
  • Smart battery management ensures the battery is always charged and thus guarantees data storage and alarm functionality in case of power failure.

 Data export / monitoring:

  • USB port for temperature data export.
  • Interfaces for external temperature monitoring such as Liebherr’s innovative SmartMonitoring: Liebherr-hosted cloud storage for maximum data security, web-based and accessible from any location worldwide, user-friendly dashboard with a multitude of highly useful additional functions, developed and maintained by Liebherr’s in-house software development team.
  • 2 x 28mm access ports for external monitoring, positioned at the top and bottom rear of the chamber to allow easy probe positioning.
Specifications

Specifications

ULT freezer
477 L
0.1°C
1 (2 compartments)
Right hand hinged
Polyurethane Foam
CE, ISO 9001, ISO 14001, ENERGYSTAR, REACH
LCD
White
Password-protected, digital with LCD display
Liebherr MediLine ULT
Manual
± 3 K
1082 x 920 x 1966 mm (42.6 x 36.2 x 77.4 in.)
10.72 ft.2
Analogue, Ethernet, Liebherr Smart Monitoring
USB, 4-20 mA analogue temperature
CO2 or LN2 Optional
120/230 V, 50 Hz, 1600 W, 10.0 A
CFC-free insulation, refrigerants GWP <6
3300 vials / kWh/24 h
26440 vials/m2
R290 / R170
280 kg
247 kg
16
16.8 cu. ft.
-40°C to -86°C
±3.0°C
2
Epoxy-enameled Steel
High-impact Powder Paint Coating
Upright
Rear fixed, front swivel/brake
Air Cooled
2 x 28 mm rear access ports
2-years parts and labour
SUFsg 5001
380 min.
605 x 606 x 1300 mm (23.8 x 23.9 x 51.2 in.)
0.995 m2</sμp
Key Lock
Independent safety controller fitted
CEE 7/7
7.9 kWh/24 h
max. 26325
2450 vials/ft2
2-stage
617 lb.
545 lb.
Boxes (352 x 50 mm)
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
View accessory products
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.