missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Urea, Ultrapure Bioreagent, J.T. Baker™
High purity reagents tested for use in biotechnology applications, such as electrophoresis, and liquid chromatography
£177.00 - £506.00
Chemical Identifiers
CAS | 57-13-6 |
---|---|
Molecular Formula | CH4N2O |
Molecular Weight (g/mol) | 60.056 |
InChI Key | XSQUKJJJFZCRTK-UHFFFAOYSA-N |
Synonym | carbamide, isourea, carbonyldiamide, ureophil, carbonyldiamine, carbamimidic acid, pseudourea, ureaphil, urevert, alphadrate |
PubChem CID | 1176 |
ChEBI | CHEBI:48376 |
IUPAC Name | urea |
SMILES | C(=O)(N)N |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
15508264
|
Avantor J.T.Baker
4111-01 |
500 g | Poly Bottle |
£177.00
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15518264
|
Avantor J.T.Baker
4111-05 |
2.5 kg | Poly Bottle |
£506.00
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
- For Protein Solubilization and Denaturation
Chemical Identifiers
57-13-6 | |
60.056 | |
carbamide, isourea, carbonyldiamide, ureophil, carbonyldiamine, carbamimidic acid, pseudourea, ureaphil, urevert, alphadrate | |
CHEBI:48376 | |
C(=O)(N)N |
CH4N2O | |
XSQUKJJJFZCRTK-UHFFFAOYSA-N | |
1176 | |
urea |
Specifications
57-13-6 | |
1L = 1.32kg | |
XSQUKJJJFZCRTK-UHFFFAOYSA-N | |
urea | |
1176 | |
60.06 | |
Urea |
CH4N2O | |
carbamide, isourea, carbonyldiamide, ureophil, carbonyldiamine, carbamimidic acid, pseudourea, ureaphil, urevert, alphadrate | |
C(=O)(N)N | |
60.056 | |
CHEBI:48376 | |
Ultrapure Bioreagent |