missing translation for 'onlineSavingsMsg'
Learn More

Lonza™ USP Reference Standard Endotoxin (RSE)

Primary standard for ensuring quality in pharmaceutical development and manufacturing. Lonza™ USP Reference Standard Endotoxin (RSE) is rigorously tested and evaluated to confirm accuracy.

Brand:  Lonza™ E700

Product Code. 11620241

  • £901.00 / Each

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

  • Contains 10,000 EU/vial
  • Stoppered and capped glass vial
  • Highly characterized
  • Use for conducting official USP-NF tests and assays
  • Analytical, clinical, pharmaceutical, and research applications
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Endotoxin reference standard
USP Reference Standard
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Lonza™ USP Reference Standard Endotoxin (RSE) > Product Type: USP reference standard

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.