missing translation for 'onlineSavingsMsg'
Learn More
Learn More
USP4 Mouse anti-Human, Mouse, Clone: 1G11G8, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech 66822-1-IG-150UL
This item is not returnable.
View return policy
Description
The protein encoded by this gene is a protease that deubiquitinates target proteins such as ADORA2A and TRIM21. The encoded protein shuttles between the nucleus and cytoplasm and is involved in maintaining operational fidelity in the endoplasmic reticulum. Three transcript variants encoding different isoforms have been found for this gene.Specifications
USP4 | |
Monoclonal | |
1 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
P35123, Q13107 | |
USP4 | |
USP4 Fusion Protein Ag28297 | |
150 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG2b |
Western Blot | |
1G11G8 | |
Unconjugated | |
USP4 | |
Deubiquitinating enzyme 4, Ubiquitin thiolesterase 4, UNP, Unph, USP4 | |
Mouse | |
Protein A | |
RUO | |
22258, 7375 | |
-20°C | |
Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction