missing translation for 'onlineSavingsMsg'
Learn More
Learn More
USP4 Mouse anti-Human, Mouse, Clone: 1G11G8, Proteintech
Mouse Monoclonal Antibody
£135.00 - £349.00
Specifications
Antigen | USP4 |
---|---|
Clone | 1G11G8 |
Concentration | 1 mg/mL |
Applications | Western Blot |
Classification | Monoclonal |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16829055
|
Proteintech
66822-1-IG-20UL |
20 μL |
£135.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
16819055
|
Proteintech
66822-1-IG-150UL |
150 μL |
£349.00
150µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
The protein encoded by this gene is a protease that deubiquitinates target proteins such as ADORA2A and TRIM21. The encoded protein shuttles between the nucleus and cytoplasm and is involved in maintaining operational fidelity in the endoplasmic reticulum. Three transcript variants encoding different isoforms have been found for this gene.Specifications
USP4 | |
1 mg/mL | |
Monoclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
Deubiquitinating enzyme 4, Ubiquitin thiolesterase 4, UNP, Unph, USP4 | |
USP4 | |
IgG2b | |
Protein A | |
Antibody |
1G11G8 | |
Western Blot | |
Unconjugated | |
Mouse | |
Mouse, Human | |
P35123, Q13107 | |
22258, 7375 | |
USP4 Fusion Protein Ag28297 | |
Primary | |
-20°C | |
USP4 |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title