missing translation for 'onlineSavingsMsg'
Learn More

UTP15 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

£135.00 - £349.00

Specifications

Antigen UTP15
Concentration 0.13 mg/mL
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
16807314
View Documents
Proteintech
17416-1-AP-20UL
20 μL
£135.00
20µL
Please to purchase this item. Need a web account? Register with us today!
16897304
View Documents
Proteintech
17416-1-AP-150UL
150 μL
£349.00
150µL
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Product No. 17416-1-AP is a rabbit polyclonal antibody raised against the full-length of human UTP15. It specifically recognizes 58 and 37 kDa isoforms.

Involved in nucleolar processing of pre-18S ribosomal RNA.
Specifications

Specifications

UTP15
Western Blot
Unconjugated
Rabbit
Human, Rat, Mouse
A2RRU3, Q8C7V3, Q8TED0
105372, 310019, 84135
UTP15 Fusion Protein Ag11161
Primary
-20°C
UTP15
0.13 mg/mL
Polyclonal
Liquid
RUO
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
NET21, UTP15
Utp15
IgG
Antigen Affinity Chromatography
Antibody
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
UTP15 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.