missing translation for 'onlineSavingsMsg'
Learn More
Learn More
UTP15 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 17416-1-AP-150UL
This item is not returnable.
View return policy
Description
Product No. 17416-1-AP is a rabbit polyclonal antibody raised against the full-length of human UTP15. It specifically recognizes 58 and 37 kDa isoforms.
Involved in nucleolar processing of pre-18S ribosomal RNA.Specifications
UTP15 | |
Polyclonal | |
Unconjugated | |
UTP15 | |
NET21, UTP15 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
105372, 310019, 84135 | |
-20°C | |
Liquid |
Western Blot | |
0.13 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
A2RRU3, Q8C7V3, Q8TED0 | |
Utp15 | |
UTP15 Fusion Protein Ag11161 | |
150 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
UTP15 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech > 150 μL; Unconjugated
Spot an opportunity for improvement?Share a Content Correction