Learn More
Cole-Parmer™ UV Tubes For Ultraviolet Equipment
Use these replacement UV tubes with ultraviolet lamps.
Brand:  Cole-Parmer™ 09815-46
 View more versions of this product
Product Code. 11820390
  • £121.00 / Each
Estimated Shipment: 10-02-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Replacement Ultraviolet Tubes
  • Product may contain trace amounts of mercury
  • Use caution with breakage, cleanup, shipping and/or disposal
Specifications
Specifications
UV Tube
6 W
15.5 mm
210.5 mm
UV-C
254 nm
6 W, 254 nm lamps