missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Wheaton™ Filtration Adapter Assembly
Makes it easier to connect standard 47 and 90mm filtration assemblies with stopper connections to a standard laboratory bottle with a GL45 thread. Wheaton™ Filtration Adapter Assembly replaces the stopper to make the connection to the bottle.
£178.00 - £815.00
Product Code | Brand | For Use With (Equipment) | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | For Use With (Equipment) | Price | Quantity & Availability | |||||
11771973
|
Wheaton™
419450 |
90 mm filtration assemblies with stopper connections |
£815.00
Each |
Estimated Shipment: 04-11-2024
Log in to see stock. |
|||||
11744179
|
Wheaton™
419505 |
47 mm filtration assemblies with stopper connections |
£178.00
Each |
Estimated Shipment: 21-11-2024
Log in to see stock. |
|||||
Description
Makes it easier to connect standard 47 and 90mm filtration assemblies with stopper connections to a standard laboratory bottle with a GL45 thread. Wheaton™ Filtration Adapter Assembly replaces the stopper to make the connection to the bottle.
- Includes a PBT hose connection for use with 1/4 in. (6mm) I.D. flexible tubing
- A GL32 PBT cap with a PTFE faced silicone sealing ring makes the connection to the filtration assembly
- A GL45 PBT cap with PTFE faced silicone sealing ring then makes the connection to the bottle.