missing translation for 'onlineSavingsMsg'
Learn More

Greiner Bio-One VACUETTE™ Multi-Drawing Blood Collection Needles

VACUETTE™ Multi-Drawing Blood Collection Needles are primarily used for routine blood collection and are intended for single-use. The needle shield is color-coded for visual recognition of the needle gauges. Needle gauges of 20G, 21G and 22G are available in lengths of 1 in. and 1.5 in.

Brand:  Greiner Bio-One 450077

 View more versions of this product

Product Code. 17790578

  • POA / Pack of 2000

Please to purchase this item. Need a web account? Register with us today!

This item requires a price update prior to being ordered.

Request a Price for this item here.

View alternative products »

Explore more special offers
This item is not returnable. View return policy

Description

Description

VACUETTE™ Multi-Drawing Blood Collection Needles are primarily used for routine blood collection and are intended for single-use. The needle shield is colour-coded for visual recognition of the needle gauges. Needle gauges of 20G, 21G and 22G are available in lengths of 1in. and 1.5in..

  • Single-use exclusively
  • Supplied in inner packs of 100 needles
  • Manufactured from stainless steel and fitted with a safety valve at one end
  • Perforated label on cap for identification and seal of integrity
  • Protective cap color indicates needle type/gauge: pink (18G), yellow (20G), green (21G), black (22G)
  • Available in lengths of 1 inch (25 mm) and 1.5 inches (38 mm)
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Multi-sample Blood Draw Needle
Sterile
Yellow
Multi-drawing Blood Collection
20 G
Yellow Protective Cap, Safety Valve at one end
3.81 cm
4°C to 25°C
Stainless Steel
CE, OSHA
Single-use
VACUETTE™ Blood Collection Tubes, Quickshield™ Safety Tube Holder
Polypropylene
1.5 in.
Synthetic Rubber
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.