missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BD Vacutainer™ Plastic Urinalysis Tubes
For the collection and transport of urine specimens. BD Vacutainer™ Plastic Urinalysis Tubes are sterile evacuated tubes with or without preservative to be used with BD collection devices. Available in many sizes with a round or conical bottom.
Brand: BD 365000
This item is not returnable.
View return policy
Description
BD Vacutainer™ urine products for the collection and transport of urine specimens offer the same level of safety and standardization as BD blood collection products.- Designed to provide accurate patient results with reduced risk of healthcare worker exposure to hazardous specimens
- Used with BD collection devices, specimen cups, 24 hour 3L containers and transfer straws
- Leak-proof urine tubes can safely transport urine specimens to the laboratory for analysis
- Options include bottom shape (round or conical) and cap style (conventional or Hemogard™)
- Most sizes are preservative/additive free
- 8mL tube (mfr. no. 364938) contains a preservative: 0.4% Chlorhexidine, 5.6% Ethyl Paraben and 94% Sodium Propionate
Specifications
Urinalysis Tube | |
Hemogard™ | |
PET | |
No additive | |
Urinalysis | |
Paper | |
9.5 mL |
Beige | |
Single-use | |
Sterile | |
Conical | |
Urine collection devices (cups, transfer straws, 24 hr. containers) | |
1000/Pk. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction