missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BD Vacutainer™ Plastic Urinalysis Tubes
For the collection and transport of urine specimens. BD Vacutainer™ Plastic Urinalysis Tubes are sterile evacuated tubes with or without preservative to be used with BD collection devices. Available in many sizes with a round or conical bottom.
£272.00 - £464.00
Specifications
Product Type | Urinalysis Tube |
---|---|
Disposable | Single-use |
Material | PET |
Sterility | Sterile |
For Use With (Application) | Urinalysis |
Product Code | Brand | Volume (Metric) | Bottom Shape | Closure Type | Price | Quantity & Availability | |||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Volume (Metric) | Bottom Shape | Closure Type | Price | Quantity & Availability | |||
12987766
|
BD
364915 |
11 mL | Round | Hemogard™ |
£342.00
Pack of 1000 |
Estimated Shipment: 21-08-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
12937766
|
BD
368500 |
4 mL | Round | Hemogard™ |
£272.00
Pack of 1000 |
Estimated Shipment: 11-09-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
12967766
|
BD
365000 |
9.5 mL | Conical | Hemogard™ |
£464.00
Pack of 1000 |
Estimated Shipment: 17-07-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
12947766
|
BD
368501 |
6 mL | Round | Hemogard™ |
£306.70
Pack of 1000 |
Estimated Shipment: 17-07-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
12957766
|
BD
364992 |
8 mL | Conical | Conventional Rubber Stopper |
£396.00
Pack of 1000 |
Estimated Shipment: 18-07-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
12977766
|
BD
364938 |
10 mL | Round | Conventional Rubber Stopper |
£431.30
Pack of 1000 |
Estimated Shipment: 17-07-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
Description
BD Vacutainer™ urine products for the collection and transport of urine specimens offer the same level of safety and standardization as BD blood collection products.- Designed to provide accurate patient results with reduced risk of healthcare worker exposure to hazardous specimens
- Used with BD collection devices, specimen cups, 24 hour 3L containers and transfer straws
- Leak-proof urine tubes can safely transport urine specimens to the laboratory for analysis
- Options include bottom shape (round or conical) and cap style (conventional or Hemogard™)
- Most sizes are preservative/additive free
- 8mL tube (mfr. no. 364938) contains a preservative: 0.4% Chlorhexidine, 5.6% Ethyl Paraben and 94% Sodium Propionate
Specifications
Urinalysis Tube | |
PET | |
Urinalysis | |
Paper |
Single-use | |
Sterile | |
Urine collection devices (cups, transfer straws, 24 hr. containers) |