missing translation for 'onlineSavingsMsg'
Learn More

BUCHI Vacuum Control for L-250

The Lyovapor™ sensors accurately track temperature and/or pressure throughout the freeze-drying process, ensuring optimized operations and facilitating accurate endpoint determination using capacitive, Pirani pressure gauges, and sample temperature probes.

Brand:  BUCHI 11080162

 View more versions of this product

Product Code. 18814171

  • £3249.15 / Each

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

  • Chose from ultimate vacuum (no control) or advanced vacuum control for process regulation and method development
Specifications

Specifications

Benchtop Freeze Dryer
0
Stainless Steel
North America
4.5 L
Ambient to -105°C
Advanced Vacuum Control
220/240 V
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers