missing translation for 'onlineSavingsMsg'
Learn More

BUCHI Vacuum Control for L-250

The Lyovapor™ sensors accurately track temperature and/or pressure throughout the freeze-drying process, ensuring optimized operations and facilitating accurate endpoint determination using capacitive, Pirani pressure gauges, and sample temperature probes.

Brand:  BUCHI 11080161

 View more versions of this product

Product Code. 30130643

  • £1740.00 / Each

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

  • Chose from ultimate vacuum (no control) or advanced vacuum control for process regulation and method development
Specifications

Specifications

Benchtop Freeze Dryer
0
Stainless Steel
North America
4.5 L
Ambient to -105°C
Ultimate Vacuum
220/240 V
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

One moment while we fetch your results.
Product Certifications
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
BUCHI Vacuum Control for L-250 > BUCHI Vacuum Control for L-250, Style; Ultimate Vacuum

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.