missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cole-Parmer™ Vacuum Filtration Holder
Optimize your vacuum filtration process with the Vacuum Filtration Holder.
Brand: Cole-Parmer™ 02920-00
This item is not returnable.
View return policy
Description
- Model 02923-20 meets EPA specifications for Hazardous Waste Toxicity Test.
- Autoclavable.
- Supplied with an aluminum clamp and a silicone rubber stopper.
- No. 5 stoppers fit standard 125 mL flasks.
- No. 8 stoppers fit 1 L flasks.
- Order no. 8B stopper 02924-02 separately to fit filter holders 02920-00 and 02921-00 to manifolds above.
- Filter holder 02923-10 is ideal for sterility testing of antibiotics and pharmaceuticals by bacteria retentive method described in USP XXI.
- This model includes funnel, filter base, aluminum clamp, and stopper.
- Model 02923-20 with sintered-glass membrane support meets EPA specifications for use in the Hazardous Waste Toxicity Test.
- Holder 02923-00, with stainless steel (SS) membrane support, provides ultraclean filtrate—screen does not shed particles.
Specifications
Autoclavable | |
Aluminum | |
Vacuum Filtration Holder |
Vacuum Filter Holder | |
1 Ea. | |
100 mL |
Product Suggestions
Customers who viewed this item also viewed.
Viewing 1-4 of
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Cole-Parmer™ Vacuum Filtration Holder > Vacuum Filtration Holder; Capacity: 100 mL
Spot an opportunity for improvement?Share a Content Correction