Learn More
Chemical Identifiers
CAS | 109-52-4 |
---|---|
Molecular Formula | C5H10O2 |
Molecular Weight (g/mol) | 102.133 |
MDL Number | MFCD00004413 |
InChI Key | NQPDZGIKBAWPEJ-UHFFFAOYSA-N |
Synonym | valeric acid, n-valeric acid, n-pentanoic acid, valerianic acid, 1-butanecarboxylic acid, propylacetic acid, butanecarboxylic acid, pentoic acid, kyselina valerova, valeric acid, n |
PubChem CID | 7991 |
ChEBI | CHEBI:17418 |
IUPAC Name | pentanoic acid |
SMILES | CCCCC(=O)O |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10123100
|
Thermo Scientific Chemicals
149570050 |
5 mL | Glass bottle |
£25.13
5mL |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10686584
|
Thermo Scientific Chemicals
149571000 |
100 mL | Glass bottle |
£30.27
100mL |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10316600
|
Thermo Scientific Chemicals
149570010 |
1 L | Glass bottle |
£45.96
1 L |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10686394
|
Thermo Scientific Chemicals
149570025 |
2.5 L | Glass bottle |
£79.60
2.50 L |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
109-52-4 | |
102.133 | |
NQPDZGIKBAWPEJ-UHFFFAOYSA-N | |
7991 | |
pentanoic acid |
C5H10O2 | |
MFCD00004413 | |
valeric acid, n-valeric acid, n-pentanoic acid, valerianic acid, 1-butanecarboxylic acid, propylacetic acid, butanecarboxylic acid, pentoic acid, kyselina valerova, valeric acid, n | |
CHEBI:17418 | |
CCCCC(=O)O |
Specifications
109-52-4 | |
185.0°C | |
99% | |
CH3(CH2)3CO2H | |
02, 299 | |
valeric acid, n-valeric acid, n-pentanoic acid, valerianic acid, 1-butanecarboxylic acid, propylacetic acid, butanecarboxylic acid, pentoic acid, kyselina valerova, valeric acid, n | |
NQPDZGIKBAWPEJ-UHFFFAOYSA-N | |
pentanoic acid | |
7991 | |
102.13 | |
0.9400g/mL | |
Authentic | |
1.4070 to 1.4100 | |
Valeric acid |
-20.0°C to +18.0°C | |
Colorless to Yellow | |
C5H10O2 | |
MFCD00004413 | |
15, 10090 | |
Solubility in water: 40g/L (20°C). Other solubilities: soluble in alcohol and ether | |
CCCCC(=O)O | |
102.133 | |
CHEBI:17418 | |
99% | |
86°C | |
Liquid | |
0.94 |
Safety and Handling
GHS H Statement
Causes severe skin burns and eye damage.
Harmful to aquatic life with long lasting effects.
GHS P Statement
IF SWALLOWED: Rinse mouth. Do NOT induce vomiting.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/shower.
Danger
- Valeric acid
Signal Word
- Danger
Hazard Category
- LONG-TERM AQUATIC HAZARD Chronic 3
- Serious eye damage/eye irritation Category 1
- Skin corrosion/irritation Category 1 B
Hazard Statement
- H314-Causes severe skin burns and eye damage.
- H318-Causes serious eye damage.
- H412-Harmful to aquatic life with long lasting effects.
Precautionary Statement
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
Supplemental information
- MIXTURE LIST-Contains : Pentanoic acid
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.