Learn More
OHAUS™ Valor™ 2000 Scale
OHAUS™ Valor™ 2000 Scale offers large LED front and rear displays, making it ideal for use in dry or washdown environments.
Brand: OHAUS™ 30072318
Code : NEW
Additional Details : Weight : 4.00000kg Transport : UN number : 2800 Chem class : 8
Description
- The sturdy polycarbonate keypad offers protection from knives and other sharp instruments used in food preparation, processing and portioning applications.
- A unique IP68 flow-thru design ensures that the NSF-certified Valor 2000 will drain moisture in the scale when used in wet conditions.
- Stabilization time is less than a half-second.
- Large front and rear LED displays allow for easy viewing so two operators can share one scale.
Specifications
50 Hours with Rechargeable Battery | |
15.00 kg | |
2.000000 g | |
V22XWE15T | |
2.000000 g | |
-10°C to +40°C | |
256 x 288 x 124 mm |
ABS housing with stainless steel platform (PW), stainless steel housing and platform (XW), up-front level indicator | |
Dual LED | |
2.000000 g | |
IPX8 | |
0.500 sec. | |
Gram, Kilogram, Ounce, Pound, Pound:Ounce | |
5.000 kg |