missing translation for 'onlineSavingsMsg'
Learn More

Vanadium carbide, 99% (metals basis), Thermo Scientific Chemicals

£48.24 - £138.69

Chemical Identifiers

CAS 12070-10-9
Molecular Formula CV
Molecular Weight (g/mol) 62.95
MDL Number MFCD00049698
InChI Key ZLANVVMKMCTKMT-UHFFFAOYSA-N
IUPAC Name methanidylidynevanadiumylium
SMILES [C-]#[V+]
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11376508
SDS View Documents
Thermo Scientific Alfa Aesar
012141.14
25 g
£48.24
25g
Please to purchase this item. Need a web account? Register with us today!
11366508
SDS View Documents
Thermo Scientific Alfa Aesar
012141.22
100 g
£138.69
100g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Vanadium carbide is used as an additive to tungsten carbide to fine the carbide crystals to improve the property of the cermets. It used in cutting tools and the steel industry. Used as material for research in electronics. Used anode materials for lithium ion batteries, and in supercapacitors.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications
Show More
Videos
Safety and Handling
Show More
SDS
Documents

Documents

Product Certifications

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Vanadium carbide, 99% (metals basis), Thermo Scientific Chemicals

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.