missing translation for 'onlineSavingsMsg'
Learn More
Vanadium(III) bromide, 99.5% (metals basis), Thermo Scientific Chemicals Product Code.: 11334929

Vanadium(III) bromide, 99.5% (metals basis), Thermo Scientific Chemicals

Product Code. 11334929
2 g
Click to view available options
Quantity:
2 g
10 g
50 g
This item is not returnable. View return policy

Product Code. 11334929

Brand: Thermo Scientific Alfa Aesar 081135.04

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Vanadium(III) bromide is used as a catalyst for C(sp3)-H fluorination reactions especially the benzylic fluorination.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 13470-26-3
Molecular Formula Br3V
Molecular Weight (g/mol) 290.654
MDL Number MFCD00049695
InChI Key ZOYIPGHJSALYPY-UHFFFAOYSA-K
Synonym vanadium bromide, vanadium tribromide, vanadium bromide vbr3, vanadium iii bromide, wln: va e3, vanadium iii bromide-v
PubChem CID 83509
IUPAC Name tribromovanadium
SMILES [V](Br)(Br)Br

Specifications

Melting Point (decomposition)
Physical Form Crystalline
Quantity 2 g
Assay Percent Range 99.5% (metals basis)
Solubility Information Soluble in water and tetrahydrofuran.
Formula Weight 290.67
Sensitivity Moisture sensitive
Density 4.00^1^8 g/mL
Chemical Name or Material Vanadium(III) bromide

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberQ29I009 Certificate TypeCertificate of Analysis Date20/02/2024 Catalog Number081135.09,081135.04,081135.18
Lot NumberU24J031 Certificate TypeCertificate of Analysis Date21/09/2023 Catalog Number081135.09,081135.04,081135.18
Lot NumberJ05W026 Certificate TypeCertificate of Analysis Date28/12/2021 Catalog Number081135.09,081135.04,081135.18
Lot NumberK30U001 Certificate TypeCertificate of Analysis Date08/12/2021 Catalog Number081135.09,081135.04,081135.18
Lot NumberU24H010 Certificate TypeCertificate of Analysis Date08/12/2021 Catalog Number081135.09,081135.04,081135.18

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Vanadium(III) bromide, 99.5% (metals basis), Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.