missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Vanadium(IV) chloride, 99+%, Thermo Scientific Chemicals
Click to view available options
Quantity:
25 g
100 g
Packaging:
Glass bottle
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications
Specifications
Boiling Point | 153.0°C |
Melting Point | -20.5°C |
Color | Brown-Red |
Physical Form | Liquid |
Quantity | 25 g |
Assay Percent Range | 99% min. (VCl4) |
Linear Formula | VCl4 |
Solubility Information | Solubility in water: vigorous reaction. Other solubilities: soluble in absolute alcohol, ether, chloroform,, acetic acid |
Formula Weight | 192.75 |
Percent Purity | ≥99% |
Show More |
Product Suggestions
Customers who viewed this item also viewed
Viewing 1-4 of
Certificates
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
5 results found
Lot Number | Certificate Type | Date | Catalog Number |
---|---|---|---|
A0396904 | Certificate of Analysis | 14/12/2022 | 299690250,299691000 |
A0394256 | Certificate of Analysis | 14/12/2022 | 299690250,299691000 |
A0401470 | Certificate of Analysis | 14/12/2022 | 299690250,299691000 |
A0436980 | Certificate of Analysis | 08/06/2022 | 299690250,299691000 |
A0425669 | Certificate of Analysis | 15/02/2022 | 299690250,299691000 |
Safety and Handling


Product Identifier
- Vanadium(IV) chloride
- Danger
- Skin corrosion/irritation Category 1 A
- Acute toxicity Category 3
- H314-Causes severe skin burns and eye damage.
- H311-Toxic in contact with skin.
- H301-Toxic if swallowed.
- H331-Toxic if inhaled.
- EUH029-Contact with water liberates toxic gas.
- P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P402+P404-Store in a dry place. Store in a closed container.
RUO – Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Vanadium(IV) chloride, 99+%, Thermo Scientific Chemicals >
Spot an opportunity for improvement?Share a Content Correction