Learn More
Vanadium(IV) oxide, 99% (metals basis), Thermo Scientific Chemicals
Description
Vanadium(IV) oxide is used in chemical sensors, energy-conserving coatings, transparent conductors and switching materials. It acts as a stationary optical shutter, optical modulators, cameras and data storage. As infrared modulators, it is used for the missile guidance systems. Thin films made up of vanadium dioxide find applications in electro-optical switches, micro-optical switch, passive smart radiators and sunshields for spacecraft.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
Melting Point | 1967°C |
Physical Form | Powder |
Quantity | 10 g |
Assay Percent Range | 99% (metals basis) |
Solubility Information | Insoluble in water. |
Formula Weight | 82.94 |
Density | 4.339 g/mL |
Chemical Name or Material | Vanadium(IV) oxide |
Safety and Handling
- Vanadium(IV) oxide
- Warning
- Serious eye damage/eye irritation Category 2
- Skin corrosion/irritation Category 2
- Specific target organ toxicity Category 3
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
- H335-May cause respiratory irritation.
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- MIXTURE LIST-Contains : Vanadium oxide
RUO – Research Use Only