missing translation for 'onlineSavingsMsg'
Learn More
Vanadium(V) oxide, 99.6% min, Thermo Scientific Chemicals Product Code.: 11306118

Vanadium(V) oxide, 99.6% min, Thermo Scientific Chemicals

Product Code. 11306118
10 g
Click to view available options
Quantity:
10 g
100 g
500 g
This item is not returnable. View return policy

Product Code. 11306118

Brand: Thermo Scientific Alfa Aesar 011094.09

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Vanadium pentoxide is the precursor to alloys of vanadium. It is widely used as an industrial catalyst for the manufacture of sulfuric acid, maleic anhydride, phthalic anhydride, adipic acid, acrylic acid, oxalic acid and anthraquinone. It finds application as a detector material in bolometers and microbolometer arrays for thermal imaging, and as an ethanol sensor in ppm (parts per million) levels. It is also used in vanadium redox batteries, which are used for energy storage in power facilities, such as wind farms. Additionally, it is used as ceramics colorant, as dye and also as a detector material in bolometers in the field of thermal imaging.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1314-62-1
Molecular Formula O5V2
Molecular Weight (g/mol) 181.88
MDL Number MFCD00011457
InChI Key GNTDGMZSJNCJKK-UHFFFAOYSA-N
Synonym vanadium v oxide, vanadium pentoxide, divanadium pentaoxide, divanadium pentoxide, vanadic anhydride, vanadin v oxide, vanadium oxide v2o5, vanadium pentaoxide, vanadic acid anhydrideShow MoreShow Less
PubChem CID 14814
ChEBI CHEBI:30045
IUPAC Name dioxovanadiooxy(dioxo)vanadium
SMILES O=[V](=O)O[V](=O)=O

Specifications

Boiling Point 1750°C (decomposition)
Melting Point 690°C
Quantity 10 g
Assay Percent Range 99.6% min
Solubility Information Slightly soluble in water.
Formula Weight 181.88
Density 3.357 g/mL
Chemical Name or Material Vanadium(V) oxide

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberY27K003 Certificate TypeCertificate of Analysis Date02/12/2024 Catalog Number011094.22,011094.36,011094.09
Lot NumberN14K003 Certificate TypeCertificate of Analysis Date22/02/2024 Catalog Number011094.22,011094.36,011094.09
Lot NumberC29Z054 Certificate TypeCertificate of Analysis Date13/10/2022 Catalog Number011094.22,011094.36,011094.09
Lot NumberT21I001 Certificate TypeCertificate of Analysis Date25/08/2022 Catalog Number011094.22,011094.36,011094.09
Lot NumberZ13H020 Certificate TypeCertificate of Analysis Date14/07/2022 Catalog Number011094.22,011094.36,011094.09

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Vanadium(V) oxide, 99.6% min, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.