missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Vanadium(V) triisopropoxide oxide, 96%, Thermo Scientific Chemicals
Click to view available options
Quantity:
1 g
5 g
25 g
100 g
500 g
Description
Vanadium(V) triisopropoxide oxide serves as a building block for a series of neutral oxovanadium complexes containing both ethoxide and acetylacetonate ligands. It is also used in catalysis. Further, it acts as a precursor and used in thermal atomic layer deposition of vanadium pentoxide.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
![TRUSTED_SUSTAINABILITY](/content/dam/fishersci/glyphs/Sustain_Badge.png)
Specifications
Boiling Point | 81°C to 83°C (2mmHg) |
Melting Point | -12°C |
Color | Orange-Yellow |
Physical Form | Liquid |
Quantity | 100 g |
Linear Formula | VO[CHO(CH3)2]3 |
UN Number | UN1993 |
Flash Point | 45°C (113°F) |
Solubility Information | Miscible with toluene,hexane and isopropanol. |
Formula Weight | 242.2 |
Show More |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'