missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cole-Parmer™ VapLock™ Solvent Delivery Caps
VapLock™ Solvent Delivery Caps reduce chemical evaporation and exposure to hazardous vapors
£53.45 - £78.30
Specifications
For Use With (Application) | Solvent Safety |
---|---|
For Use With (Equipment) | Solvent-Compatible Containers |
Thread Size (English) | 1/4 in.-28 UNF(F) |
Product Code | Brand | Type | Material | Size | Price | Quantity & Availability | |||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Type | Material | Size | Price | Quantity & Availability | |||
15814524
|
Cole-Parmer™
12018-04 |
Three 1/4 in.-28 ports | Blue/white PTFE, PP | GL45 |
£68.20
Each |
In Stock
Log in to see stock. |
|||
15824524
|
Cole-Parmer™
12018-01 |
304 SS Port Thread Inserts, three 1/4 in.-28 | Blue/white PTFE, PP, SS | GL45 |
£78.30
Each |
Estimated Shipment: 12-12-2024
Log in to see stock. |
|||
15854474
|
Cole-Parmer™
12018-54 |
Two 1/4 in.-28 ports | Blue/white PTFE, PP | GL25 |
£53.45
Each |
In Stock
Log in to see stock. |
|||
Description
Use with semi-rigid tubing equal to or less than 1/8 in (3.2 mm) ODWarranty
None
Specifications
Solvent Safety | |
Solvent-Compatible Containers | |
1/4 in.-28 UNF(F) |