Learn More
Emma Safety Footwear Vera Safety Shoes, Red
An easy shoe for wipe clean hygiene. The upper is fully washable unique microfiber and features Hydro-Tec™ Sanitized Silver for dry, fresh feet. The protective toe cap prevents staining and is lubricant and fat resistant. Developed for use on hard tiled floors and other easy-to-clean floors.
Brand:  Emma Safety Footwear MM50652438
Additional Details : Weight : 1.26300kg
 View more versions of this product
Product Code. 15978511
  • £143.10 / 1 pair
Estimated Shipment: 30-07-2024
to see stock.
Add to basket
Description
Description
  • S2-Safety shoe for wet working conditions
  • Microfiber construction
  • GripForce™ Duo Heat Resistant Outsole
  • Chrome-free lining
  • Sanitized
Specifications
Specifications
EN ISO 20345:2011
White
Hydro-Tec™ Sanitized Silver
38
Elastic
Vera, Red
Microfiber
Steel