Learn More
Emma Safety Footwear Vera Safety Shoes, Red
An easy shoe for wipe clean hygiene. The upper is fully washable unique microfiber and features Hydro-Tec™ Sanitized Silver for dry, fresh feet. The protective toe cap prevents staining and is lubricant and fat resistant. Developed for use on hard tiled floors and other easy-to-clean floors.
£143.10 - £166.00
Specifications
Certifications/Compliance | EN ISO 20345:2011 |
---|---|
Closure Type | Elastic |
Color | White |
Design | Vera, Red |
Liner Material | Hydro-Tec™ Sanitized Silver |
Product Code | Brand | Size | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Size | Price | Quantity & Availability | |||||
15968511
|
Emma Safety Footwear
MM50652437 |
37 |
£166.00
1 pair |
Estimated Shipment: 19-08-2024
Log in to see stock. |
|||||
15978511
|
Emma Safety Footwear
MM50652438 |
38 |
£143.10
1 pair |
Estimated Shipment: 30-07-2024
Log in to see stock. |
|||||
15988511
|
Emma Safety Footwear
MM50652439 |
39 |
£143.10
1 pair |
Estimated Shipment: 30-07-2024
Log in to see stock. |
|||||
15998511
|
Emma Safety Footwear
MM50652440 |
40 |
£143.10
1 pair |
Estimated Shipment: 30-07-2024
Log in to see stock. |
|||||
15908521
|
Emma Safety Footwear
MM50652441 |
41 |
£143.10
1 pair |
Estimated Shipment: 19-08-2024
Log in to see stock. |
|||||
15918521
|
Emma Safety Footwear
MM50652442 |
42 |
£143.10
1 pair |
Estimated Shipment: 15-08-2024
Log in to see stock. |
|||||
15928521
|
Emma Safety Footwear
MM50652443 |
43 |
£143.10
1 pair |
Estimated Shipment: 22-07-2024
Log in to see stock. |
|||||
15938521
|
Emma Safety Footwear
MM50652444 |
44 |
£143.10
1 pair |
Estimated Shipment: 19-08-2024
Log in to see stock. |
|||||
15948521
|
Emma Safety Footwear
MM50652445 |
45 |
£159.00
1 pair |
Estimated Shipment: 19-08-2024
Log in to see stock. |
|||||
15958521
|
Emma Safety Footwear
MM50652446 |
46 |
£143.10
1 pair |
Estimated Shipment: 04-09-2024
Log in to see stock. |
|||||
15968521
|
Emma Safety Footwear
MM50652447 |
47 |
£143.10
1 pair |
Estimated Shipment: 22-08-2024
Log in to see stock. |
|||||
15978521
|
Emma Safety Footwear
MM50652448 |
48 |
£143.10
1 pair |
Estimated Shipment: 19-08-2024
Log in to see stock. |
|||||
15988521
|
Emma Safety Footwear
MM50652449 |
49 |
£159.00
1 pair |
Estimated Shipment: 22-08-2024
Log in to see stock. |
|||||
Description
- S2-Safety shoe for wet working conditions
- Microfiber construction
- GripForce™ Duo Heat Resistant Outsole
- Chrome-free lining
- Sanitized
Specifications
EN ISO 20345:2011 | |
White | |
Hydro-Tec™ Sanitized Silver | |
Steel |
Elastic | |
Vera, Red | |
Microfiber |