Learn More
Cytiva (formerly Pall Lab) Versapor™ Acrylic Copolymer 1200 Membrane Disc Filter
Disc Filters useful for prefiltration of particle-laden samples, serum filtration, and dissolution testing.
Brand:  Cytiva (formerly Pall Lab) 66393
Additional Details : Weight : 0.11400kg
 View more versions of this product
Product Code. 17134561
  • £114.55 / Pack of 100
Estimated Shipment: 30-08-2024
to see stock.
Add to basket
Description
Description
  • Protects instrumentation against particulate build up
  • This product is not approved for use in medical, clinical, surgical or other patient applications
Specifications
Specifications
Versapor (Acrylic Copolymer on a Non-woven Support)
Non-sterile
25 mm
1.2 μm
94 μm