missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cytiva (formerly Pall Lab) Versapor™ Acrylic Copolymer 1200 Membrane Disc Filter
Disc Filters useful for prefiltration of particle-laden samples, serum filtration, and dissolution testing.
£114.55 - £570.00
Specifications
Material (Membrane) | Versapor (Acrylic Copolymer on a Non-woven Support) |
---|---|
Pore Size | 1.2 μm |
Sterility | Non-sterile |
Thickness (Metric) | 94 μm |
Product Code | Brand | Diameter (Metric) Membrane | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Diameter (Metric) Membrane | Price | Quantity & Availability | |||||
17134561
|
Cytiva (formerly Pall Lab)
66393 |
25 mm |
£114.55
Pack of 100 |
Estimated Shipment: 30-08-2024
Log in to see stock. |
|||||
17144561
|
Cytiva (formerly Pall Lab)
66394 |
47 mm |
£167.00
Pack of 100 |
Estimated Shipment: 30-08-2024
Log in to see stock. |
|||||
17164561
|
Cytiva (formerly Pall Lab)
66397 |
293 mm |
£570.00
Pack of 25 |
Estimated Shipment: 30-08-2024
Log in to see stock. |
|||||
17154561
|
Cytiva (formerly Pall Lab)
66396 |
142 mm |
£286.00
Pack of 25 |
Estimated Shipment: 30-08-2024
Log in to see stock. |
|||||
Description
- Protects instrumentation against particulate build up
- This product is not approved for use in medical, clinical, surgical or other patient applications
Specifications
Versapor (Acrylic Copolymer on a Non-woven Support) | |
1.2 μm | |
Non-sterile | |
94 μm |