Learn More
Scientific Industries SI™ Vertical Microtube Holder
Requires no additional attachment for use. Scientific Industries SI™ Vertical Microtube Holder snaps microtubes into place.
Brand:  Scientific Industries SI™ SI-V209
 View more versions of this product
Product Code. 15587365
  • £125.00 / Each
Estimated Shipment: 10-04-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Holds tubes vertically
  • Snaps on to any Vortex-Genie 2 Mixer and the Vortex-Genie Pulse
  • Optimized for high speed vortexing a small number of tubes
  • No additional attachment necessary
Specifications
Specifications
Vortex-Genie 2 Mixer and Vortex-Genie Pulse
9
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
One moment while we fetch your results.
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
Scientific Industries SI™ Vertical Microtube Holder > Holds: 9 Tubes