Learn More
Cole-Parmer™ Vial Holder for HG-600 Geno/Grinder™ 2010 Homogenizer
This Vial Holder for HG-600 Geno/Grinder™ 2010 Homogenizer securely hold vials in place while processing samples.
£206.00
Specifications
Product Type Holder
For Use With (Equipment) HG-600 Geno/Grinder™ 2010 Homogenizer
Material Foam
Packaging Quantity 1 Ea.
View More Specs
Products 1
Product Code Brand Length (Metric) Holds Price Quantity & Availability  
Product Code Brand Length (Metric) Holds Price Quantity & Availability  
12517336
View Documents
Cole-Parmer™
2197
11.9 cm 24 Standard 15 mL Conical Bottom Centrifuge Tubes
£206.00
Each
In Stock
to see stock.
Add to basket
Description
Description
  • Safely grind samples in the HG-600 Geno/Grinder™ 2010.
  • Vial holders are suitable for use with the HG-600 Geno/Grinder™ 2010 Homogenizer.
  • Order homogenizer and vials separately.
Specifications
Specifications
Holder
HG-600 Geno/Grinder™ 2010 Homogenizer
Foam
1 Ea.