Learn More
Invitrogen™ ViewRNA™ Cell Plus Assay Kit
The ViewRNA Cell Plus Assay is a novel assay that combines immunocytochemistry with ViewRNA technology, a proprietary fluorescent in situ hybridization and sequential branched-DNA amplification technique, to visualize both RNA with single-molecule sensitivity and protein in individual cells.
Brand:  Invitrogen™ 88-19000-99
Product Code. 16318755
  • £1322.00 / 1 set
Explore more special offers
Description
Description
This assay enables simultaneous detection of up to three RNA targets in combination with immunophenotyping for cell surface or intracellular proteins using both indirect and direct immunocytochemistry to allow for detailed characterization of specific cell subpopulations. For simultaneous detection of a fourth RNA target, the ViewRNA ISH Cell 740 Module (Cat. No. QVC0200) can be combined with this kit. It allows analysis of an additional target in the 740 channel (AlexaFluor 750).
This ViewRNA Cell Plus Assay kit contains all the reagents needed to conduct the assay. Target Probe sets for genes of interest and antibodies are sold separately.
Reactivity/species
Probes can be designed for any species. These are the species most frequently used/tested: Armenian hamster, baboon, bovine, canine, cat, chicken, chimpanzee, cow, cynomolgus monkey, donkey, goat, golden Syrian hamster, guinea pig, hamster, horse, human, macaque, monkey, mouse, non-human primate, olive baboon, pig, pigtailed macaque, rabbit, rat, rhesus monkey, sheep
Reported application
Microscopy, immunocytochemistry
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Primer-probe Detection
Microscopy, Immunocytochemistry
1 kit
Kit
RNA In Situ Hybridization Assay Kit
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
5 results found
Lot Number Certificate Type Date Catalog Number
Lot Number415951-204 Certificate TypeCertificate of Analysis Date10/04/2025 Catalog Number
Lot Number415951-305 Certificate TypeCertificate of Analysis Date10/04/2025 Catalog Number
Lot Number415951-304 Certificate TypeCertificate of Analysis Date10/04/2025 Catalog Number
Lot Number415951-104 Certificate TypeCertificate of Analysis Date10/04/2025 Catalog Number
Lot Number415951-105 Certificate TypeCertificate of Analysis Date10/04/2025 Catalog Number
Special Offers
Special Offers
Product Content Correction
Product Title
Invitrogen™ ViewRNA™ Cell Plus Assay Kit > Primer-Probe Detection