Learn More
Tocris Bioscience™ Vilazodone hydrochloride
Low molecular weight, organic compound that may help regulate biological process.
Brand:  Tocris Bioscience™ 6202/50
96.76 GBP valid until 2024-12-16
Use promo code "21630" to get your promotional price.
 View more versions of this product
Product Code. 18734868
  • £125.00 / 50mg
Explore more special offers
Description
Description
Vilazodone hydrochloride is a potent 5-HT1A partial agonist and serotonin-selective reuptake inhibitor (SSRI) (IC50 values are 0.2 and 0.5 nM respectively). Antidepressant.
Specifications
Specifications
Agonist
163521-08-2
477.99g/mol
5-HT1A Receptor Antagonists
5-HT1A
C26H27N5O2.HCl
Store at -20°C