missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Vimentin Mouse anti-Human, Mouse, Rat, Clone: 3H9D1, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech 60330-1-IG-20UL
This item is not returnable.
View return policy
Description
Vimentin is a member of the intermediate filament family. Intermediate filamentents, along with microtubules and actin microfilaments, make up the cytoskeleton. The protein is responsible for maintaining cell shape, integrity of the cytoplasm, and stabilizing cytoskeletal interactions. It is also involved in the immune response, and controls the transport of low-density lipoprotein (LDL)-derived cholesterol from a lysosome to the site of esterification. It functions as an organizer of a number of critical proteins involved in attachment, migration, and cell signaling. Mutations in this gene causes a domit, pulverulent cataract.Specifications
Vimentin | |
Monoclonal | |
0.8 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
P08670, P20152, P31000 | |
VIM | |
Vimentin Fusion Protein Ag0489 | |
20 μL | |
Primary | |
Mouse, Human, Rat | |
Antibody | |
IgG1 |
Flow Cytometry, Immunocytochemistry, Western Blot, Immunofluorescence, Immunohistochemistry (Paraffin) | |
3H9D1 | |
Unconjugated | |
VIM | |
VIM, Vimentin | |
Mouse | |
Protein G | |
RUO | |
22352, 7431, 81818 | |
-20°C | |
Liquid |
Product Suggestions
Customers who viewed this item also viewed
Viewing 1-4 of
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Vimentin Mouse anti-Human, Mouse, Rat, Clone: 3H9D1, Proteintech > 20 μL; Unconjugated
Spot an opportunity for improvement?Share a Content Correction