Learn More
HC Innovations Vinco-SanWipe Cleaning & Sanitising Facilities Anti-Bac and Anti-Viral Wipe
Formulated for use in gyms, airports, workplaces, facilities management, industrial areas, showrooms and offices
Brand: HC Innovations CP128
Additional Details : CAS Number : 7173-51-5 Weight : 3.60000kg
Description
- Can be used on hands and surfaces, for best results use 1 wipe to remove soiling and the second wipe to sanitize.
- These facilities wipes are best used on hands and hard surfaces such as desks and equipment in offices, door handles, grab handles, machinery, keyboards, washroom areas and other areas where there is high foot fall.
Specifications
Sanitizing Wipes | |
25 gsm | |
Store in a Dry Place | |
Angled | |
Cleaning and sanitizing | |
1-ply | |
Non-sterile | |
Recyclable |
Not Autoclavable | |
White | |
Yes | |
200 x 200 mm | |
150 Wipes | |
14.3 Chemical / 85.7 Water | |
Fabric, Chemical | |
Keep upright |