missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Brady™ Vinyl Continuous Tape
B-7569 Vinyl Continuous Tape is ideal for indoor/outdoor facility labels, including 5S labels, safety labels, warning labels, pipe markers, equipment labels and visual workplace labels.
Brand: Brady™ 120922
This item is not returnable.
View return policy
Description
- B-7569 is ideal for indoor/outdoor facility labels, including 5S labels, safety labels, warning labels, pipe markers, equipment labels and visual workplace labels.
Specifications
BS5609:1986 Section 2 and section 3 | |
Continuous Vinyl Tape for BBP3X/S3XXX/i3300 Printers | |
White | |
Vinyl | |
Labels | |
BBP30, BBP31, BBP33, BBP35, BBP37, i3300, S3000, S3100, i5300 | |
1 | |
1 Roll/Box | |
Brady Workstation, LabelMark, MarkWare | |
-20°C to +75°C | |
0.102 mm | |
98 ft. | |
2.244 in. |
B30C-2250-7569-WT | |
Permanent Acrylic, Permanent | |
Gloss | |
Thermal Transfer | |
Facility Identification, Floor marking, Hazard communication Identification, Pipe and valve marking, Safety Identification, Warehouse marking | |
5S labels, safety labels, warning labels, pipe markers, equipment labels and visual workplace labels | |
-20°C | |
1 Ea. | |
-4°F to +167°F | |
0.004 in. | |
30 m x 57 mm | |
30 m | |
57 mm |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction