missing translation for 'onlineSavingsMsg'
Learn More

Vinyltrimethylsilane, 97%, Thermo Scientific Chemicals

£52.46 - £150.75

Chemical Identifiers

CAS 754-05-2
Molecular Formula C5H12Si
Molecular Weight (g/mol) 100.24
MDL Number MFCD00008606
InChI Key GCSJLQSCSDMKTP-UHFFFAOYSA-N
Synonym vinyltrimethylsilane, trimethyl vinyl silane, trimethylvinylsilane, silane, ethenyltrimethyl, trimethylsilyl ethylene, vinyl trimethylsilane, silane, trimethylvinyl, vinyltrimethysilane, trimethylsilyl ethene, ethenyl-trimethylsilane
PubChem CID 79102
IUPAC Name ethenyl(trimethyl)silane
SMILES C[Si](C)(C)C=C
View More Specs

Products 2
Product Code Brand Quantity Packaging Price Quantity & Availability  
Product Code Brand Quantity Packaging Price Quantity & Availability  
10635272
View Documents
Thermo Scientific Acros
200330250
25 mL Glass bottle
£52.46
25mL
Please to purchase this item. Need a web account? Register with us today!
10645272
View Documents
Thermo Scientific Acros
200331000
100 mL Glass bottle
£150.75
100mL
Please to purchase this item. Need a web account? Register with us today!
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

754-05-2
100.24
GCSJLQSCSDMKTP-UHFFFAOYSA-N
79102
C[Si](C)(C)C=C
C5H12Si
MFCD00008606
vinyltrimethylsilane, trimethyl vinyl silane, trimethylvinylsilane, silane, ethenyltrimethyl, trimethylsilyl ethylene, vinyl trimethylsilane, silane, trimethylvinyl, vinyltrimethysilane, trimethylsilyl ethene, ethenyl-trimethylsilane
ethenyl(trimethyl)silane

Specifications

754-05-2
0.6800g/mL
-24°C
96% min. (GC)
H2C=CHSi(CH3)3
MFCD00008606
0.68
GCSJLQSCSDMKTP-UHFFFAOYSA-N
ethenyl(trimethyl)silane
79102
97%
Vinyltrimethylsilane, 97%
Colorless
55°C
Authentic
C5H12Si
1.3895 to 1.3915
09,498; 10,444; 12,566; 13,343
vinyltrimethylsilane, trimethyl vinyl silane, trimethylvinylsilane, silane, ethenyltrimethyl, trimethylsilyl ethylene, vinyl trimethylsilane, silane, trimethylvinyl, vinyltrimethysilane, trimethylsilyl ethene, ethenyl-trimethylsilane
C[Si](C)(C)C=C
100.24
100.24
Liquid
Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Highly flammable liquid and vapor.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Ground/Bond container and receiving equipment.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger

missing translation for 'einecsNumber' : 212-042-9

missing translation for 'tsca' : TSCA

Product Identifier
  • Vinyltrimethylsilane

Signal Word
  • Danger

Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Flammable liquid Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3

Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H225-Highly flammable liquid and vapour.

Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P240-Ground/bond container and receiving equipment.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

SDS
Documents

Documents

Certificates

RUO – Research Use Only