Learn More
Gibco™ Viral Production Medium
Gibco Viral Production Medium is a chemically defined, serum-free, and protein-free medium. It is animal origin-free and specially developed for growth and transfection of Viral Production Cells 2.0 (suspension-adapted HEK 293F-derived cells).
£210.00 - £1692.00
Specifications
Cell Line | Viral Production Cells 2.0 |
---|---|
Cell Type | HEK 293 cells |
Classification | Serum-free, Animal Origin-free, Chemically-defined, Protein-free |
Form | Liquid |
Product Type | AAV Production Medium |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
17371889
|
Gibco™
A4817901 |
1 L |
£210.00
1 L |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
17651536
|
Gibco™
A4817902 |
6 L |
£1106.00
Pack of 6 |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
17381889
|
Gibco™
A4817903 |
10 L |
£1692.00
10 L |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
- A chemically defined, serum-free, protein-free, and animal origin-free medium
- Specially developed for growth and transfection of Viral Production Cells 2.0 (suspension-adapted HEK 293F-derived cells)
- For adeno-associated virus (AAV) production as part of the AAV-MAX Helper-Free AAV Production System
- Supports growth of suspension Viral Production Cells 2.0 to densities of over 1.2 x 107 cells/mL
- Enables production of high AAV titers, greater than 5 x 1010 vg/mL (unpurified)
- Supports growth and transfection of Viral Production Cells 2.0 from shake flask to bioreactor scales
- Viral Production Medium requires supplementation with 4 mM GlutaMAX Supplement (Cat. No. 35050038) prior to use
Specifications
Viral Production Cells 2.0 | |
Serum-free, Animal Origin-free, Chemically-defined, Protein-free | |
AAV Production Medium | |
Glucose, Sodium Pyruvate, Sodium Pyruvate, GlutaMAX | |
AAV Production | |
Room Temperature | |
RUO - Research Use Only | |
Mammalian |
HEK 293 cells | |
Liquid | |
Sterile | |
No Glutamine | |
AAV-MAX | |
7 | |
Suspension Cell Culture |
Safety and Handling
missing translation for 'shelfLife' : 12 months from date of manufacture
For Research Use Only. Not for use in diagnostic procedures.
Your input is important to us. Please complete this form to provide feedback related to the content on this product.