Learn More
Waters Corporation Viridis BEH 2-EP Column
Designed and tested for SFC, the Viridis columns demonstrate a wide range of selectivity for greater resolution.
Brand:  Waters Corporation 186005766
Encompass
 View more versions of this product
Product Code. 17123292
  • £5814.00 / Each
Explore more special offers
Description
Description
  • The reduction and control of surface silanol activity on Viridis Hybrid particles delivers, under SFC conditions, excellent peak shapes – without the use of mobile-phase additives – even for well-retained basic achiral compounds.
Specifications
Specifications
30 mm
6000 psi (415 Bar)
5 μm
1/Ea.
Column
50 mm
Spherical
130 Å
No
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Waters Corporation Viridis BEH 2-EP Column