Learn More
Buerkle™ ViscoSampler
ViscoSampler with powerful suction for viscous media. The media is drawn up and discharged using the suction flask. Specially developed for viscous substances such as slurries, silt, wet clay and soil samples, soil sediments, oils, emulsions, creams etc.
Brand: Buerkle™ 5331-2060
374.80 GBP valid until 2024-12-31
Use promo code "23267" to get your promotional price.
Code : A0
Additional Details : Weight : 0.54350kg
Description
- Powerful suction for viscous media
- The media is drawn up and discharged using the suction flask
- For use with slurries, silt, wet clay and soil samples, soil sediments, oils, emulsions, creams etc
- Easy to clean
- All surfaces are poreless and without crevices
- Dirt accumulation is prevented
- Only round screw threads tried and tested in the field of food hygiene are used
- Disassembly and cleaning are extremely simple
Specifications
PTFE, FEP | |
160 mL | |
60 cm |