missing translation for 'onlineSavingsMsg'
Learn More

Sampling Systems™ Viscous Sampler

Operates like a large syringe by having the operator pull the handle up to suck the product into the sampler. Sampling Systems™ Viscous Sampler is ideal for sampling creams, shampoos, soaps, honey and other medium viscous materials.

Brand:  Sampling Systems™ 7000T-600

 View more versions of this product

Product Code. 15889381

  • £591.00 / Each
Estimated Shipment: 30-01-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • Suitable for medium viscosity materials.
  • Manufactured from PP, 316L Stainless Steel or PTFE
  • Works like a large a syringe
  • The operator pulls up the handle in order to suck the product into the sampler
Specifications

Specifications

Viscous Fluid Sampler
160 mL
25 mm
PTFE
Medium Viscosity Materials
600 mm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.