missing translation for 'onlineSavingsMsg'
Learn More

VistaLab™ Technologies Reagent Reservoirs
GREENER_CHOICE

Unique trough-within-trough design maximizes reagent recovery

Brand:  VistaLab™ Technologies 3054-1012

 View more versions of this product

Product Code. 15742962

  • £91.90 / Pack of 100
Estimated Shipment: 28-03-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • Polystyrene with trimmed base corners
  • Distinct pour-off spouts to empty reservoirs without spills
  • Well-marked graduated increments on inside wall minimize reagent waste
  • Wide base and sturdy construction provide stability and help to avoid spills
  • Molded corner structures allow easy separation of bulk packaged reservoirs
  • Available in 25, 50, 100mL and 1.8L volumes; 25mL available with divided configuation for multichannel pipetting of two reagents simultaneously
  • 100mL model (Cat. No. 21-385-111) is supplied with lid
  • Available individually wrapped or bulk packaged, gamma sterilized or nonsterile

Order Info

Reservoir lid sold separately (Cat. No. 21-385-110).

Specifications

Specifications

Pyrogen-free
10 mL
Sterile
7.5 in.
Bulk
7 in.
White
Polystyrene
Reagent Reservoir
13 in.
100/Cs.
Single-use
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.