Learn More
VistaLab™ Technologies Reagent Reservoirs
GREENER_CHOICE
Unique trough-within-trough design maximizes reagent recovery
£79.80 - £236.00
Specifications
Certifications/Compliance Pyrogen-free
Type Reagent Reservoir
For Use With (Application) General Liquid Handling
Autoclavable Not Autoclavable
Channels 1
View More Specs
Products 24
Product Code Brand Capacity (Metric) Sterility Packaging Quantity Price Quantity & Availability  
Product Code Brand Capacity (Metric) Sterility Packaging Quantity Price Quantity & Availability  
11915732
GREENER_CHOICE
View Documents
VistaLab™ Technologies
3054-1000
100 mL Sterile Bulk 10/bag x 10; 100/Cs.
£91.90
Pack of 100
Estimated Shipment: 23-05-2025
to see stock.
Add to basket
11925732
GREENER_CHOICE
View Documents
VistaLab™ Technologies
3054-1001
100 mL Sterile Individually Wrapped 50/Cs.
£137.00
Pack of 50
Estimated Shipment: 23-05-2025
to see stock.
Add to basket
11945732
View Documents
VistaLab™ Technologies
3054-1004
25 mL (divided) Sterile Bulk 10/bag x 10; 100/Cs.
£91.90
Pack of 100
Estimated Shipment: 27-05-2025
to see stock.
Add to basket
11925742
GREENER_CHOICE
View Documents
VistaLab™ Technologies
3054-3001
100 mL (with lid) Sterile Individually Wrapped 50/Cs.
£236.00
Pack of 50
Estimated Shipment: 28-07-2025
to see stock.
Add to basket
11955732
GREENER_CHOICE
View Documents
VistaLab™ Technologies
3054-1003
25 mL Sterile Individually Wrapped 50/Cs.
£127.00
Pack of 50
Estimated Shipment: 11-07-2025
to see stock.
Add to basket
11975732
View Documents
VistaLab™ Technologies
3054-1006
50 mL Sterile Bulk 10/bag x 10; 100/Cs.
£120.00
Pack of 100
Estimated Shipment: 06-06-2025
to see stock.
Add to basket
11985732
View Documents
VistaLab™ Technologies
3054-1007
50 mL Sterile Individually Wrapped 50/Cs.
£98.95
Pack of 50
Estimated Shipment: 21-05-2025
to see stock.
Add to basket
11935722
GREENER_CHOICE
View Documents
VistaLab™ Technologies
3054-2001
100 mL Sterile Individually Wrapped 100/Cs.
£196.00
Pack of 100
Estimated Shipment: 14-08-2025
to see stock.
Add to basket
11995732
GREENER_CHOICE
View Documents
VistaLab™ Technologies
3054-2000
100 mL Sterile Bulk 20/bag x 10; 200/Cs.
£172.00
Pack of 200
Estimated Shipment: 19-06-2025
to see stock.
Add to basket
11965722
GREENER_CHOICE
View Documents
VistaLab™ Technologies
3054-2006
50 mL Sterile Bulk 20/bag x 10; 200/Cs.
£164.00
Pack of 200
Estimated Shipment: 22-05-2025
to see stock.
Add to basket
11945722
GREENER_CHOICE
View Documents
VistaLab™ Technologies
3054-2002
25 mL Sterile Bulk 20/bag x 10; 200/Cs.
£169.00
Pack of 200
Estimated Shipment: 29-05-2025
to see stock.
Add to basket
11905742
GREENER_CHOICE
View Documents
VistaLab™ Technologies
3054-2007
50 mL Sterile Individually Wrapped 100/Cs.
£173.00
Pack of 100
Estimated Shipment: 01-07-2025
to see stock.
Add to basket
11955722
GREENER_CHOICE
View Documents
VistaLab™ Technologies
3054-2003
25 mL Sterile Individually Wrapped 100/Cs.
£173.00
Pack of 100
In Stock
to see stock.
Edge
Add to basket
11975742
View Documents
VistaLab™ Technologies
4054-1002
25 mL Non-sterile Bulk 10/bag x 10; 100/Cs.
£79.80
Pack of 100
Estimated Shipment: 27-05-2025
to see stock.
Add to basket
11985742
GREENER_CHOICE
View Documents
VistaLab™ Technologies
4054-1004
25 mL (divided) Non-sterile Bulk 10/bag x 10; 100/Cs.
£80.80
Pack of 100
Estimated Shipment: 28-07-2025
to see stock.
Add to basket
11915752
GREENER_CHOICE
View Documents
VistaLab™ Technologies
4054-1006
50 mL Non-sterile Bulk 10/bag x 10; 100/Cs.
£79.80
Pack of 100
Estimated Shipment: 29-05-2025
to see stock.
Add to basket
11965742
GREENER_CHOICE
View Documents
VistaLab™ Technologies
4054-1000
100 mL Non-sterile Bulk 10/bag x 10; 100/Cs.
£83.85
Pack of 100
Estimated Shipment: 14-08-2025
to see stock.
Add to basket
11935732
View Documents
VistaLab™ Technologies
3054-1002
25 mL Sterile Bulk 10/bag x 10; 100/Cs.
£117.00
Pack of 100
In Stock
to see stock.
Edge
Add to basket
11965732
GREENER_CHOICE
View Documents
VistaLab™ Technologies
3054-1005
25 mL (divided) Sterile Individually Wrapped 50/Cs.
£98.95
Pack of 50
Estimated Shipment: 21-05-2025
to see stock.
Add to basket
15742962
GREENER_CHOICE
View Documents
VistaLab™ Technologies
3054-1012
10 mL Sterile Bulk 100/Cs.
£91.90
Pack of 100
Estimated Shipment: 06-08-2025
to see stock.
Add to basket
16445094
GREENER_CHOICE
View Documents
VistaLab™ Technologies
3054-1013
10 mL Sterile Individually packed 50/Cs.
£98.95
Pack of 50
Estimated Shipment: 21-05-2025
to see stock.
Add to basket
16455094
GREENER_CHOICE
View Documents
VistaLab™ Technologies
3054-2012
10 mL Sterile Bulk 200 in. 20 bags of 10
£205.00
Pack of 200
Estimated Shipment: 21-05-2025
to see stock.
Add to basket
16465094
GREENER_CHOICE
View Documents
VistaLab™ Technologies
3054-2013
10 mL Sterile Individually packed 100/Cs.
£173.00
Pack of 100
In Stock
to see stock.
Edge
Add to basket
16405104
GREENER_CHOICE
View Documents
VistaLab™ Technologies
4054-1012
10 mL Non-sterile Bulk 100/Cs.
£90.90
Pack of 100
Estimated Shipment: 29-05-2025
to see stock.
Add to basket
Description
Description
  • Polystyrene with trimmed base corners
  • Distinct pour-off spouts to empty reservoirs without spills
  • Well-marked graduated increments on inside wall minimize reagent waste
  • Wide base and sturdy construction provide stability and help to avoid spills
  • Molded corner structures allow easy separation of bulk packaged reservoirs
  • Available in 25, 50, 100mL and 1.8L volumes; 25mL available with divided configuation for multichannel pipetting of two reagents simultaneously
  • 100mL model (Cat. No. 21-385-111) is supplied with lid
  • Available individually wrapped or bulk packaged, gamma sterilized or nonsterile
Order Info
Reservoir lid sold separately (Cat. No. 21-385-110).
Specifications
Specifications
Pyrogen-free
General Liquid Handling
1
V-bottom
Reagent Reservoir
Not Autoclavable
Single-use
Videos
SDS
Documents
Documents
Product Certifications
Product Content Correction
Product Title
VistaLab™ Technologies Reagent Reservoirs