missing translation for 'onlineSavingsMsg'
Learn More

VistaLab™ Technologies Reagent Reservoirs
GREENER_CHOICE

Unique trough-within-trough design maximizes reagent recovery

Brand:  VistaLab™ Technologies 3054-2013

 View more versions of this product

Product Code. 16465094

  • £173.00 / Pack of 100
Estimated Shipment: 13-01-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • Polystyrene with trimmed base corners
  • Distinct pour-off spouts to empty reservoirs without spills
  • Well-marked graduated increments on inside wall minimize reagent waste
  • Wide base and sturdy construction provide stability and help to avoid spills
  • Molded corner structures allow easy separation of bulk packaged reservoirs
  • Available in 25, 50, 100mL and 1.8L volumes; 25mL available with divided configuation for multichannel pipetting of two reagents simultaneously
  • 100mL model (Cat. No. 21-385-111) is supplied with lid
  • Available individually wrapped or bulk packaged, gamma sterilized or nonsterile

Order Info

Reservoir lid sold separately (Cat. No. 21-385-110).

Specifications

Specifications

Pyrogen-free
10 mL
Sterile
12.5 in.
Individually packed
12 in.
White
Polystyrene
Reagent Reservoir
12 in.
100/Cs.
Single-use
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers