Learn More
Emma Safety Footwear Vito S3 Shoe
Maintain that sharp look, even when wearing your safety gear with these Emma Vito safety shoes.
Brand:  Emma Safety Footwear MM11400048
 View more versions of this product
Product Code. 17562446
  • £169.00 / 1 pair
Estimated Shipment: 14-01-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Smart work shoes with water-resistant full-grain and nubuck leather uppers
  • Hydro-Tec antibacterial and Wellness leather lining
  • Antibacterial inlays with leather top layers
  • Steel toecaps and aramid fibre penetration-resistant midsoles
  • Polyurethane (PU) comfort midsoles with thermoplastic polyurethane (TPU) outsoles
  • Regular fit
  • Worn shoes can be returned to the manufacturer for recycling (postage provided); keep receipt and contact us for more details when the time comes
  • Outsoles are resistant to fuel oil
  • Excellent slip resistance
  • Energy-absorbing heels for comfort and support
  • Antistatic
Specifications
Specifications
ISO 20345:2011 S3 FO
Black
Leather
Steel
Low-cut
Laces
Government, Local Authority, Oil and Gas
48
Extra-wide Width
Safety Shoe