missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ismatec™ Viton™ Tubing for Peristaltic Pumps
Has excellent chemical resistance, especially to acids and alkalis, making it an excellent choice for demanding applications such as acid and solvent transfer. It also handles temperatures from 8 to 400°F, making it ideal for high-temperature work.
Brand: Ismatec™ 97632-42
This item is not returnable.
View return policy
Description
- Tubing undergoes robust testing to provide optimal performance/pump life hours with peristaltic pump equipment
- Precision extruded to tight tolerances for high repeatability, accuracy, and reproducibility
- Global market leader in range of formulations for most extensive chemical compatibility
- Regulatory approvals to match your unique application
Specifications
FDA approved; Prop 65 NA (no Prop 65 chemicals) | |
Viton | |
15.2 m | |
0°C | |
45 mL/min | |
25.3 psi, 1.75 bar | |
Microbore Tubing |
2.06 mm | |
50 ft. | |
C/L Pump Series | |
42 | |
3/32 in. | |
150°C |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction