missing translation for 'onlineSavingsMsg'
Learn More

Ismatec™ Viton™ Tubing for Peristaltic Pumps

Has excellent chemical resistance, especially to acids and alkalis, making it an excellent choice for demanding applications such as acid and solvent transfer. It also handles temperatures from 8 to 400°F, making it ideal for high-temperature work.

Brand:  Ismatec™ 97632-48

 View more versions of this product

Product Code. 15384698

  • £638.00 / 50ft
Estimated Shipment: 20-02-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • Tubing undergoes robust testing to provide optimal performance/pump life hours with peristaltic pump equipment
  • Precision extruded to tight tolerances for high repeatability, accuracy, and reproducibility
  • Global market leader in range of formulations for most extensive chemical compatibility
  • Regulatory approvals to match your unique application
Specifications

Specifications

FDA approved; Prop 65 NA (no Prop 65 chemicals)
Viton
15.2 m
0°C
66 mL/min
25.3 psi, 1.75 bar
Microbore Tubing
2.79 mm
50 ft.
C/L Pump Series
48
1/8 in.
150°C
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.